Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759023.4 | XP_002759069.2 | 2 - 3 | 397 | 81 | |
| XM_008982817.2 | XP_008981065.1 | 2 - 3 | 407 | 81 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map