Assay Details
| Gene Symbol: SLC41A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758713.4 | XP_002758759.1 | 5 - 6 | 705 | 65 | |
| XM_008982295.2 | XP_008980543.1 | 5 - 6 | 698 | 65 | ||
| XM_017965160.1 | XP_017820649.1 | 5 - 6 | 734 | 65 | ||
More Information
Set Membership: |
|

SLC41A3
Genomic Map