Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974445.1 | XP_017829934.1 | 5 - 6 | 5827 | 61 | |
| XM_017974446.1 | XP_017829935.1 | 5 - 6 | 5708 | 61 | ||
| XM_017974447.1 | XP_017829936.1 | 5 - 6 | 5813 | 61 | ||
| XM_017974448.1 | XP_017829937.1 | 4 - 5 | 6002 | 61 | ||
| XM_017974449.1 | XP_017829938.1 | 3 - 4 | 8061 | 61 | ||
| XM_017974450.1 | XP_017829939.1 | 5 - 6 | 5668 | 61 | ||
| XM_017974452.1 | XP_017829941.1 | 5 - 6 | 5549 | 61 | ||
| XM_017974453.1 | XP_017829942.1 | 4 - 5 | 5486 | 61 | ||
More Information
Set Membership: |
|

AHDC1
Genomic Map