Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746534.3 | XP_002746580.1 | 3 - 4 | 522 | 83 | |
| XM_017971084.1 | XP_017826573.1 | 3 - 4 | 547 | 83 | ||
| XM_017971085.1 | XP_017826574.1 | 4 - 5 | 659 | 83 | ||
More Information
Set Membership: |
|

MED20
Genomic Map