Assay Details
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759301.4 | XP_002759347.1 | 3 - 4 | 1744 | 97 | |
XM_008983357.2 | XP_008981605.1 | 3 - 4 | 1767 | 97 | ||
XM_008983358.2 | XP_008981606.1 | 3 - 4 | 1783 | 97 | ||
XM_017965633.1 | XP_017821122.1 | 3 - 4 | 1524 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
