Assay Details
Gene Symbol: TPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017977942.1 | XP_017833431.1 | 9 - 10 | 1128 | 95 | |
XM_017977943.1 | XP_017833432.1 | 8 - 9 | 1977 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
