Assay Details
| Gene Symbol: ZNF827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992631.2 | XP_008990879.1 | 3 - 4 | 1860 | 67 | |
| XM_017970216.1 | XP_017825705.1 | 3 - 4 | 1861 | 67 | ||
| XM_017970217.1 | XP_017825706.1 | 3 - 4 | 1801 | 67 | ||
More Information
Set Membership: |
|

ZNF827
Genomic Map