Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199929.1 | NP_001186858.1 | 3 - 4 | 459 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SELENOT| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199929.1 | NP_001186858.1 | 3 - 4 | 459 | 96 | |