Assay Details
Gene Symbol: YIPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757868.4 | XP_002757914.1 | 3 - 4 | 671 | 122 | |
XM_017964428.1 | XP_017819917.1 | 3 - 4 | 917 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
