Assay Details
| Gene Symbol: YAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754639.4 | XP_002754685.1 | 8 - 9 | 1644 | 63 | |
| XM_002754640.4 | XP_002754686.1 | 7 - 8 | 1596 | 63 | ||
| XM_003734140.3 | XP_003734188.1 | 7 - 8 | 1530 | 63 | ||
| XM_009007178.2 | XP_009005426.1 | 8 - 9 | 1414 | 63 | ||
| XM_009007179.2 | XP_009005427.1 | 8 - 9 | 1656 | 63 | ||
| XM_009007180.2 | XP_009005428.1 | 8 - 9 | 1402 | 63 | ||
| XM_009007181.2 | XP_009005429.1 | 7 - 8 | 1366 | 63 | ||
| XM_009007182.2 | XP_009005430.1 | 7 - 8 | 1608 | 63 | ||
| XM_009007183.2 | XP_009005431.1 | 7 - 8 | 1354 | 63 | ||
| XM_009007184.2 | XP_009005432.1 | 7 - 8 | 1542 | 63 | ||
| XM_009007185.2 | XP_009005433.1 | 6 - 7 | 1494 | 63 | ||
| XM_009007186.2 | XP_009005434.1 | 6 - 7 | 1482 | 63 | ||
More Information
Set Membership: |
|

YAP1
Genomic Map