Assay Details
| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752755.3 | XP_002752801.1 | 6 - 7 | 1289 | 90 | |
| XM_009004200.2 | XP_009002448.1 | 6 - 7 | 1425 | 90 | ||
| XM_009004201.2 | XP_009002449.1 | 6 - 7 | 1420 | 90 | ||
| XM_009004202.2 | XP_009002450.1 | 6 - 7 | 1289 | 90 | ||
| XM_009004203.2 | XP_009002451.1 | 5 - 6 | 1333 | 90 | ||
| XM_009004204.2 | XP_009002452.1 | 6 - 7 | 1427 | 90 | ||
More Information
Set Membership: |
|

RAB3IP
Genomic Map