Assay Details
| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001969.1 | XP_009000217.1 | 6 - 7 | 1639 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAN1A2| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001969.1 | XP_009000217.1 | 6 - 7 | 1639 | 60 | |
Set Membership: |
|