Assay Details
| Gene Symbol: RNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760258.4 | XP_002760304.1 | 4 - 5 | 799 | 85 | |
| XM_008984832.2 | XP_008983080.1 | 4 - 5 | 565 | 85 | ||
| XM_008984833.2 | XP_008983081.1 | 4 - 5 | 619 | 85 | ||
More Information
Set Membership: |
|

RNF2
Genomic Map