Assay Details
| Gene Symbol: ASXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747306.2 | XP_002747352.1 | 12 - 13 | 2447 | 89 | |
| XM_008995553.2 | XP_008993801.1 | 11 - 12 | 1898 | 89 | ||
| XM_017971854.1 | XP_017827343.1 | 12 - 13 | 1927 | 89 | ||
| XM_017971855.1 | XP_017827344.1 | 11 - 12 | 1924 | 89 | ||
| XM_017971856.1 | XP_017827345.1 | 11 - 12 | 1774 | 89 | ||
More Information
Set Membership: |
|

ASXL1
Genomic Map