Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760689.3 | XP_002760735.1 | 2 - 3 | 848 | 117 | |
| XM_008985480.1 | XP_008983728.1 | 2 - 3 | 848 | 117 | ||
More Information
Set Membership: |
|

ETNK2
Genomic Map