Assay Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007609.2 | XP_009005857.2 | 3 - 4 | 1302 | 76 | |
| XM_009007610.2 | XP_009005858.2 | 3 - 4 | 1301 | 76 | ||
| XM_009007611.2 | XP_009005859.2 | 3 - 4 | 1301 | 76 | ||
More Information
Set Membership: |
|

SYT9
Genomic Map