Assay Details
| Gene Symbol: PPP1R3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005776.2 | XP_009004024.1 | 1 - 2 | 903 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3E| Gene Symbol: PPP1R3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005776.2 | XP_009004024.1 | 1 - 2 | 903 | 79 | |
Set Membership: |
|