Assay Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745954.4 | XP_002746000.1 | 5 - 6 | 802 | 97 | |
| XM_008993537.2 | XP_008991785.1 | 6 - 7 | 1481 | 97 | ||
| XM_008993538.2 | XP_008991786.1 | 5 - 6 | 1418 | 97 | ||
| XM_008993539.2 | XP_008991787.1 | 5 - 6 | 2066 | 97 | ||
| XM_008993542.2 | XP_008991790.1 | 5 - 6 | 782 | 97 | ||
| XM_017970647.1 | XP_017826136.1 | 5 - 6 | 672 | 97 | ||
| XM_017970648.1 | XP_017826137.1 | 7 - 8 | 1568 | 97 | ||
| XM_017970649.1 | XP_017826138.1 | 4 - 5 | 594 | 97 | ||
| XM_017970650.1 | XP_017826139.1 | 5 - 6 | 785 | 97 | ||
| XM_017970651.1 | XP_017826140.1 | 5 - 6 | 782 | 97 | ||
More Information
Set Membership: |
|

LCORL
Genomic Map