Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748079.4 | XP_002748125.1 | 1 - 2 | 256 | 103 | |
| XM_008996785.1 | XP_008995033.1 | 1 - 2 | 256 | 103 | ||
More Information
Set Membership: |
|

CCDC43
Genomic Map