Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744164.3 | XP_002744210.1 | 2 - 3 | 770 | 78 | |
| XM_017967677.1 | XP_017823166.1 | 2 - 3 | 571 | 78 | ||
| GenBank mRNA | EF215213.1 | - | 1 - 2 | 54 | 78 | |
More Information
Set Membership: |
|

CAMLG
Genomic Map