Assay Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994094.2 | XP_008992342.1 | 5 - 6 | 995 | 86 | |
| XM_008994095.2 | XP_008992343.1 | 5 - 6 | 1005 | 86 | ||
| XM_017971606.1 | XP_017827095.1 | 5 - 6 | 1003 | 86 | ||
| XM_017971607.1 | XP_017827096.1 | 4 - 5 | 1926 | 86 | ||
More Information
Set Membership: |
|

TRIM39
Genomic Map