Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997221.2 | XP_008995469.1 | 2 - 3 | 188 | 76 | |
| XM_017972577.1 | XP_017828066.1 | 1 - 2 | 939 | 76 | ||
| XM_017972578.1 | XP_017828067.1 | 1 - 2 | 939 | 76 | ||
| XM_017972579.1 | XP_017828068.1 | 1 - 2 | 939 | 76 | ||
More Information
Set Membership: |
|

LYRM9
Genomic Map