Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747541.4 | XP_002747587.1 | 1 - 2 | 1990 | 76 | |
| XM_008995906.2 | XP_008994154.1 | 1 - 2 | 1998 | 76 | ||
More Information
Set Membership: |
|

NNAT
Genomic Map