Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981333.2 | XP_008979581.1 | 555 | 87 | ||
| XM_017970840.1 | XP_017826329.1 | 553 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCHL3| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981333.2 | XP_008979581.1 | 555 | 87 | ||
| XM_017970840.1 | XP_017826329.1 | 553 | 87 | |||
Set Membership: |
|