Assay Details
Gene Symbol: LOC100390879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749792.2 | XP_002749838.1 | 3 - 4 | 584 | 65 | |
XM_008999492.2 | XP_008997740.1 | 2 - 3 | 1439 | 65 | ||
XM_008999494.2 | XP_008997742.1 | 4 - 5 | 624 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
