Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744379.4 | XP_002744425.2 | 2 - 3 | 1454 | 69 | |
| XM_008986989.2 | XP_008985237.1 | 2 - 3 | 1399 | 69 | ||
More Information
Set Membership: |
|

KCTD16
Genomic Map