Assay Details
| Gene Symbol: MRPS18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746309.3 | XP_002746355.2 | 4 - 5 | 756 | 64 | |
| XM_017971612.1 | XP_017827101.1 | 4 - 5 | 756 | 64 | ||
More Information
Set Membership: |
|

MRPS18B
Genomic Map