Assay Details
| Gene Symbol: PPP1R14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755469.4 | XP_002755515.1 | 3 - 4 | 565 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R14B| Gene Symbol: PPP1R14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755469.4 | XP_002755515.1 | 3 - 4 | 565 | 56 | |
Set Membership: |
|