Assay Details
| Gene Symbol: PDCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757424.4 | XP_002757470.2 | 4 - 5 | 519 | 76 | |
| XM_017964099.1 | XP_017819588.1 | 3 - 4 | 274 | 76 | ||
More Information
Set Membership: |
|

PDCL3
Genomic Map