Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756447.3 | XP_002756493.1 | 8 | 1881 | 99 | |
| XM_017979143.1 | XP_017834632.1 | 7 | 1809 | 99 | ||
| GenBank mRNA | EF215199.1 | - | 1 | 219 | 99 | |
More Information
Set Membership: |
|

LGI1
Genomic Map