Assay Details
| Gene Symbol: TPMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746189.4 | XP_002746235.1 | 5 - 6 | 573 | 79 | |
| XM_008993903.2 | XP_008992151.1 | 4 - 5 | 480 | 79 | ||
More Information
Set Membership: |
|

TPMT
Genomic Map