Assay Details
| Gene Symbol: LOC103787289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008361.2 | XP_009006609.1 | 4 - 5 | 558 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103787289| Gene Symbol: LOC103787289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008361.2 | XP_009006609.1 | 4 - 5 | 558 | 75 | |
Set Membership: |
|