Assay Details
| Gene Symbol: MRPL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752851.4 | XP_002752897.1 | 5 - 6 | 583 | 69 | |
| XM_009004365.2 | XP_009002613.1 | 6 - 7 | 681 | 69 | ||
| XM_009004367.2 | XP_009002615.1 | 6 - 7 | 684 | 69 | ||
| XM_017976278.1 | XP_017831767.1 | 5 - 6 | 599 | 69 | ||
More Information
Set Membership: |
|

MRPL42
Genomic Map