Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003146.2 | XP_009001394.1 | 5 - 6 | 635 | 81 | |
| XM_009003147.2 | XP_009001395.1 | 5 - 6 | 593 | 81 | ||
| XM_009003148.2 | XP_009001396.1 | 5 - 6 | 624 | 81 | ||
| XM_009003150.2 | XP_009001398.1 | 4 - 5 | 500 | 81 | ||
| XM_009003153.2 | XP_009001401.1 | 3 - 4 | 331 | 81 | ||
| XM_009003154.2 | XP_009001402.1 | 5 - 6 | 634 | 81 | ||
| XM_009003155.2 | XP_009001403.1 | 5 - 6 | 633 | 81 | ||
| XM_009003156.2 | XP_009001404.1 | 5 - 6 | 633 | 81 | ||
More Information
Set Membership: |
|

ASB15
Genomic Map