Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756870.4 | XP_002756916.1 | 4 - 5 | 472 | 65 | |
| XM_008979121.2 | XP_008977369.1 | 4 - 5 | 702 | 65 | ||
| XM_008979122.2 | XP_008977370.1 | 4 - 5 | 455 | 65 | ||
| XM_008979124.2 | XP_008977372.1 | 4 - 5 | 521 | 65 | ||
| XM_008979125.2 | XP_008977373.1 | 4 - 5 | 446 | 65 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map