Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735640.2 | XP_003735688.1 | 4 - 5 | 816 | 111 | |
| XM_008988459.2 | XP_008986707.1 | 4 - 5 | 565 | 111 | ||
| XM_008988460.1 | XP_008986708.1 | 4 - 5 | 790 | 111 | ||
| XM_008988461.2 | XP_008986709.1 | 3 - 4 | 446 | 111 | ||
| XM_008988462.1 | XP_008986710.1 | 3 - 4 | 670 | 111 | ||
More Information
Set Membership: |
|

JOSD2
Genomic Map