Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748261.4 | XP_002748307.1 | 3 - 4 | 827 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFAIP1| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748261.4 | XP_002748307.1 | 3 - 4 | 827 | 60 | |
Set Membership: |
|