Assay Details
Gene Symbol: LOC100390879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749792.2 | XP_002749838.1 | 6 - 7 | 1018 | 70 | |
XM_008999492.2 | XP_008997740.1 | 5 - 6 | 1873 | 70 | ||
XM_008999494.2 | XP_008997742.1 | 7 - 8 | 1058 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
