Assay Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744086.3 | XP_002744132.2 | 5 - 6 | 601 | 92 | |
XM_017966969.1 | XP_017822458.1 | 4 - 5 | 819 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
