Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999048.2 | XP_008997296.1 | 3 - 4 | 620 | 78 | |
| XM_008999050.2 | XP_008997298.1 | 4 - 5 | 787 | 78 | ||
| XM_008999051.2 | XP_008997299.1 | 5 - 6 | 907 | 78 | ||
| XM_008999052.2 | XP_008997300.1 | 4 - 5 | 731 | 78 | ||
| XM_008999053.2 | XP_008997301.1 | 3 - 4 | 337 | 78 | ||
| XM_008999056.2 | XP_008997304.1 | 3 - 4 | 620 | 78 | ||
| XM_017973546.1 | XP_017829035.1 | 2 - 3 | 320 | 78 | ||
More Information
Set Membership: |
|

TFPI
Genomic Map