Assay Details
| Gene Symbol: COLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981192.2 | XP_008979440.1 | 4 - 5 | 640 | 71 | |
| XM_008981193.2 | XP_008979441.1 | 4 - 5 | 467 | 71 | ||
More Information
Set Membership: |
|

COLEC11
Genomic Map