Assay Details
| Gene Symbol: APITD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204928.1 | NP_001191857.1 | 1 - 2 | 455 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APITD1| Gene Symbol: APITD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204928.1 | NP_001191857.1 | 1 - 2 | 455 | 68 | |
Set Membership: |
|