Assay Details
| Gene Symbol: APITD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204928.1 | NP_001191857.1 | 2 - 3 | 582 | 95 | |
| XM_009000308.2 | XP_008998556.1 | 2 - 3 | 407 | 95 | ||
More Information
Set Membership: |
|

APITD1
Genomic Map