Assay Details
| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752969.4 | XP_002753015.3 | 6 - 7 | 2189 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUAK1| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752969.4 | XP_002753015.3 | 6 - 7 | 2189 | 75 | |
Set Membership: |
|