Assay Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980482.1 | XP_008978730.1 | 552 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR54| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980482.1 | XP_008978730.1 | 552 | 78 | ||