Assay Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761323.3 | XP_002761369.1 | 4 - 5 | 771 | 91 | |
| XM_008986506.2 | XP_008984754.1 | 3 - 4 | 467 | 91 | ||
More Information
Set Membership: |
|

BTG3
Genomic Map