Assay Details
Gene Symbol: CALHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756422.3 | XP_002756468.1 | 4 | 1847 | 98 | |
XM_017979281.1 | XP_017834770.1 | 3 | 2304 | 98 | ||
XM_017979282.1 | XP_017834771.1 | 3 | 1729 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
