Assay Details
| Gene Symbol: NFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967745.1 | XP_017823234.1 | 7 - 8 | 1348 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFIC| Gene Symbol: NFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967745.1 | XP_017823234.1 | 7 - 8 | 1348 | 56 | |
Set Membership: |
|