Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756841.3 | XP_002756887.2 | 9 - 10 | 2192 | 76 | |
| XM_017963692.1 | XP_017819181.1 | 10 - 11 | 2419 | 76 | ||
| XM_017963693.1 | XP_017819182.1 | 11 - 12 | 2306 | 76 | ||
More Information
Set Membership: |
|

RHOBTB2
Genomic Map