Assay Details
| Gene Symbol: WNT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733706.2 | XP_003733754.2 | 2 - 3 | 305 | 101 | |
| XM_017975858.1 | XP_017831347.1 | 1 - 2 | 1115 | 101 | ||
More Information
Set Membership: |
|

WNT5B
Genomic Map